missing translation for 'onlineSavingsMsg'
Learn More

Trimethoxy[3-(phenylamino)propyl]silane 98.0+%, TCI America™

Catalog No. p-7135145
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P145825G 25 g
P1458250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P145825G Supplier TCI America Supplier No. P145825G
Only null left

Chemical Identifiers

CAS 3068-76-6
Molecular Formula C12H21NO3Si
Molecular Weight (g/mol) 255.389
MDL Number MFCD00053673
InChI Key KBJFYLLAMSZSOG-UHFFFAOYSA-N
Synonym n-3-trimethoxysilyl propyl aniline, trimethoxy 3-phenylamino propyl silane, n-phenylaminopropyltrimethoxysilane, unii-7au1h1fv6a, benzenamine, n-3-trimethoxysilyl propyl, n-phenyl-3-aminopropyltrimethoxysilane, 7au1h1fv6a, n-3-trimethoxysilylpropyl aniline, 3-phenylamino propyltrimethoxysilane
PubChem CID 76476
IUPAC Name N-(3-trimethoxysilylpropyl)aniline
SMILES CO[Si](CCCNC1=CC=CC=C1)(OC)OC

Specifications

Color Yellow
Boiling Point 310°C
Quantity 25 g
UN Number 3267
Formula Weight 255.39
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Trimethoxy[3-(phenylamino)propyl]silane
EINECSNumber (3)-2644
TSCA Yes