CAS RN 3068-76-6
IUPAC Name:
N-[3-(trimethoxysilyl)propyl]aniline
Synonyms:
N-[3-(trimethoxysilyl)propyl]aniline
Molecular Weight (g/mol):
255.39
Molecular Formula:
C12H21NO3Si
InChi Key:
KBJFYLLAMSZSOG-UHFFFAOYSA-N
SMILES:
CO[Si](CCCNC1=CC=CC=C1)(OC)OC
1
–
1
of
1
results
1
Promotions Available
3-(Phenylamino)propyltrimethoxysilane, 96%
3-(Phenylamino)propyltrimethoxysilane, 96%
CAS: 3068-76-6 Molecular Formula: C12H21NO3Si Molecular Weight (g/mol): 255.389 MDL Number: MFCD00053673 InChI Key: KBJFYLLAMSZSOG-UHFFFAOYSA-N Synonym: n-3-trimethoxysilyl propyl aniline,trimethoxy 3-phenylamino propyl silane,n-phenylaminopropyltrimethoxysilane,unii-7au1h1fv6a,benzenamine, n-3-trimethoxysilyl propyl,n-phenyl-3-aminopropyltrimethoxysilane,7au1h1fv6a,n-3-trimethoxysilylpropyl aniline,3-phenylamino propyltrimethoxysilane PubChem CID: 76476 IUPAC Name: N-(3-trimethoxysilylpropyl)aniline SMILES: CO[Si](CCCNC1=CC=CC=C1)(OC)OC