missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethoxy[3-(methylamino)propyl]silane 95.0+%, TCI America™
Supplier: TCI America T2868100G
Specifications
| Trimethoxy[3-(methylamino)propyl]silane | |
| Yellow | |
| C7H19NO3Si | |
| 100 g | |
| DVYVMJLSUSGYMH-UHFFFAOYSA-N | |
| methyl[3-(trimethoxysilyl)propyl]amine | |
| 76480 | |
| ≥95.0% (GC,T) |
| 3069-25-8 | |
| 106°C | |
| MFCD00048021 | |
| n-methylaminopropyltrimethoxysilane, n-methyl-3-trimethoxysilyl propan-1-amine, trimethoxy 3-methylamino propyl silane, 1-propanamine, n-methyl-3-trimethoxysilyl, n-methyl-3-trimethoxysilyl propylamine, n-methyl-3-aminopropyltrimethoxysilane, 3-methylaminopropyl-trimethoxysilane, methyl 3-trimethoxysilyl propyl amine, n-methyl-3-aminopropyl-trimethoxysilane, acmc-20ak27 | |
| CNCCC[Si](OC)(OC)OC | |
| 193.32 | |
| 193.32 | |
| Liquid |
Chemical Identifiers
| 3069-25-8 | |
| 193.32 | |
| DVYVMJLSUSGYMH-UHFFFAOYSA-N | |
| 76480 | |
| CNCCC[Si](OC)(OC)OC |
| C7H19NO3Si | |
| MFCD00048021 | |
| n-methylaminopropyltrimethoxysilane, n-methyl-3-trimethoxysilyl propan-1-amine, trimethoxy 3-methylamino propyl silane, 1-propanamine, n-methyl-3-trimethoxysilyl, n-methyl-3-trimethoxysilyl propylamine, n-methyl-3-aminopropyltrimethoxysilane, 3-methylaminopropyl-trimethoxysilane, methyl 3-trimethoxysilyl propyl amine, n-methyl-3-aminopropyl-trimethoxysilane, acmc-20ak27 | |
| methyl[3-(trimethoxysilyl)propyl]amine |
Safety and Handling
TSCA : Yes