missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethoxy[3-(methylamino)propyl]silane 95.0+%, TCI America™
$145.40 - $423.14
Chemical Identifiers
| CAS | 3069-25-8 |
|---|---|
| Molecular Formula | C7H19NO3Si |
| Molecular Weight (g/mol) | 193.32 |
| MDL Number | MFCD00048021 |
| InChI Key | DVYVMJLSUSGYMH-UHFFFAOYSA-N |
| Synonym | n-methylaminopropyltrimethoxysilane, n-methyl-3-trimethoxysilyl propan-1-amine, trimethoxy 3-methylamino propyl silane, 1-propanamine, n-methyl-3-trimethoxysilyl, n-methyl-3-trimethoxysilyl propylamine, n-methyl-3-aminopropyltrimethoxysilane, 3-methylaminopropyl-trimethoxysilane, methyl 3-trimethoxysilyl propyl amine, n-methyl-3-aminopropyl-trimethoxysilane, acmc-20ak27 |
| PubChem CID | 76480 |
| IUPAC Name | methyl[3-(trimethoxysilyl)propyl]amine |
| SMILES | CNCCC[Si](OC)(OC)OC |
Chemical Identifiers
| 3069-25-8 | |
| 193.32 | |
| DVYVMJLSUSGYMH-UHFFFAOYSA-N | |
| 76480 | |
| CNCCC[Si](OC)(OC)OC |
| C7H19NO3Si | |
| MFCD00048021 | |
| n-methylaminopropyltrimethoxysilane, n-methyl-3-trimethoxysilyl propan-1-amine, trimethoxy 3-methylamino propyl silane, 1-propanamine, n-methyl-3-trimethoxysilyl, n-methyl-3-trimethoxysilyl propylamine, n-methyl-3-aminopropyltrimethoxysilane, 3-methylaminopropyl-trimethoxysilane, methyl 3-trimethoxysilyl propyl amine, n-methyl-3-aminopropyl-trimethoxysilane, acmc-20ak27 | |
| methyl[3-(trimethoxysilyl)propyl]amine |
Specifications
| 3069-25-8 | |
| 106°C | |
| MFCD00048021 | |
| n-methylaminopropyltrimethoxysilane, n-methyl-3-trimethoxysilyl propan-1-amine, trimethoxy 3-methylamino propyl silane, 1-propanamine, n-methyl-3-trimethoxysilyl, n-methyl-3-trimethoxysilyl propylamine, n-methyl-3-aminopropyltrimethoxysilane, 3-methylaminopropyl-trimethoxysilane, methyl 3-trimethoxysilyl propyl amine, n-methyl-3-aminopropyl-trimethoxysilane, acmc-20ak27 | |
| CNCCC[Si](OC)(OC)OC | |
| 193.32 | |
| 193.32 | |
| Liquid |
| Yellow | |
| C7H19NO3Si | |
| 25 g | |
| DVYVMJLSUSGYMH-UHFFFAOYSA-N | |
| methyl[3-(trimethoxysilyl)propyl]amine | |
| 76480 | |
| ≥95.0% (GC,T) | |
| Trimethoxy[3-(methylamino)propyl]silane |
Safety and Handling
TSCA : Yes