accessibility menu, dialog, popup

UserName

CH3HNSiOH3COH3COH3C

CAS RN 3069-25-8

IUPAC Name: methyl[3-(trimethoxysilyl)propyl]amine
Synonyms: methyl[3-(trimethoxysilyl)propyl]amine
Molecular Weight (g/mol): 193.32
Molecular Formula: C7H19NO3Si
InChi Key: DVYVMJLSUSGYMH-UHFFFAOYSA-N
SMILES: CNCCC[Si](OC)(OC)OC
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results