missing translation for 'onlineSavingsMsg'
Learn More

Trimethoxy(3,3,3-trifluoropropyl)silane 98.0+%, TCI America™

Supplier:  TCI America T272025G

 View more versions of this product

Catalog No. T272025G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Trimethoxy(3,3,3-trifluoropropyl)silane
Colorless
C6H13F3O3Si
25 g
trimethoxy 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl trimethoxysilane, 3,3,3-trifluoropropyltrimethoxysilane, silane, trimethoxy 3,3,3-trifluoropropyl, 1,1,1-trifluoro-3-trimethoxysilyl propane, silane, trimethoxy trifluoropropyl, silane,trimethoxy 3,3,3-trifluoropropyl, acmc-1ajbz, fas-3, cf3 ch2 2si och3 3
CO[Si](CCC(F)(F)F)(OC)OC
218.25
218.25
Liquid
429-60-7
144°C
MFCD00039266
1993
JLGNHOJUQFHYEZ-UHFFFAOYSA-N
trimethoxy(3,3,3-trifluoropropyl)silane
67933
≥98.0% (GC)

Chemical Identifiers

429-60-7
218.25
JLGNHOJUQFHYEZ-UHFFFAOYSA-N
67933
CO[Si](CCC(F)(F)F)(OC)OC
C6H13F3O3Si
MFCD00039266
trimethoxy 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl trimethoxysilane, 3,3,3-trifluoropropyltrimethoxysilane, silane, trimethoxy 3,3,3-trifluoropropyl, 1,1,1-trifluoro-3-trimethoxysilyl propane, silane, trimethoxy trifluoropropyl, silane,trimethoxy 3,3,3-trifluoropropyl, acmc-1ajbz, fas-3, cf3 ch2 2si och3 3
trimethoxy(3,3,3-trifluoropropyl)silane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2079

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions