accessibility menu, dialog, popup

UserName

CH3OSiFFFOH3COH3C

CAS RN 429-60-7

IUPAC Name: trimethoxy(3,3,3-trifluoropropyl)silane
Synonyms: trimethoxy(3,3,3-trifluoropropyl)silane
Molecular Weight (g/mol): 218.25
Molecular Formula: C6H13F3O3Si
InChi Key: JLGNHOJUQFHYEZ-UHFFFAOYSA-N
SMILES: CO[Si](CCC(F)(F)F)(OC)OC
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results