missing translation for 'onlineSavingsMsg'
Learn More

Trimethoxy(3,3,3-trifluoropropyl)silane 98.0+%, TCI America™

Catalog No. p-7137371
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27205G 5 g
T272025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27205G Supplier TCI America Supplier No. T27205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 429-60-7
Molecular Formula C6H13F3O3Si
Molecular Weight (g/mol) 218.25
MDL Number MFCD00039266
InChI Key JLGNHOJUQFHYEZ-UHFFFAOYSA-N
Synonym trimethoxy 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl trimethoxysilane, 3,3,3-trifluoropropyltrimethoxysilane, silane, trimethoxy 3,3,3-trifluoropropyl, 1,1,1-trifluoro-3-trimethoxysilyl propane, silane, trimethoxy trifluoropropyl, silane,trimethoxy 3,3,3-trifluoropropyl, acmc-1ajbz, fas-3, cf3 ch2 2si och3 3
PubChem CID 67933
IUPAC Name trimethoxy(3,3,3-trifluoropropyl)silane
SMILES CO[Si](CCC(F)(F)F)(OC)OC

Specifications

Color Colorless
Boiling Point 144°C
Quantity 5 g
UN Number 1993
Formula Weight 218.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethoxy(3,3,3-trifluoropropyl)silane
EINECSNumber (2)-2079
TSCA Yes