missing translation for 'onlineSavingsMsg'
Learn More

Trimethoxy(1H,1H,2H,2H-heptadecafluorodecyl)silane 98.0+%, TCI America™

Supplier:  TCI America T291725G

 View more versions of this product

Catalog No. T291725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Trimethoxy(1H,1H,2H,2H-heptadecafluorodecyl)silane
Colorless
C13H13F17O3Si
25 g
HJIMAFKWSKZMBK-UHFFFAOYSA-N
568.30
Liquid
83048-65-1
80°C
MFCD07368748
Trimethoxy(1H,1H,2H,2H-perfluorodecyl)silane, 1H,1H,2H,2H-Heptadecafluorodecyltrimethoxysilane
568.30
≥98.0% (GC)

Chemical Identifiers

83048-65-1
568.30
HJIMAFKWSKZMBK-UHFFFAOYSA-N
C13H13F17O3Si
MFCD07368748
Trimethoxy(1H,1H,2H,2H-perfluorodecyl)silane, 1H,1H,2H,2H-Heptadecafluorodecyltrimethoxysilane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions