missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethoxy(1H,1H,2H,2H-heptadecafluorodecyl)silane 98.0+%, TCI America™
$322.11 - $1071.83
Chemical Identifiers
| CAS | 83048-65-1 |
|---|---|
| Molecular Formula | C13H13F17O3Si |
| Molecular Weight (g/mol) | 568.30 |
| MDL Number | MFCD07368748 |
| InChI Key | HJIMAFKWSKZMBK-UHFFFAOYSA-N |
| Synonym | Trimethoxy(1H,1H,2H,2H-perfluorodecyl)silane, 1H,1H,2H,2H-Heptadecafluorodecyltrimethoxysilane |
Chemical Identifiers
| 83048-65-1 | |
| 568.30 | |
| HJIMAFKWSKZMBK-UHFFFAOYSA-N |
| C13H13F17O3Si | |
| MFCD07368748 | |
| Trimethoxy(1H,1H,2H,2H-perfluorodecyl)silane, 1H,1H,2H,2H-Heptadecafluorodecyltrimethoxysilane |
Specifications
| 83048-65-1 | |
| 80°C | |
| MFCD07368748 | |
| Trimethoxy(1H,1H,2H,2H-perfluorodecyl)silane, 1H,1H,2H,2H-Heptadecafluorodecyltrimethoxysilane | |
| 568.30 | |
| ≥98.0% (GC) | |
| Trimethoxy(1H,1H,2H,2H-heptadecafluorodecyl)silane |
| Colorless | |
| C13H13F17O3Si | |
| 5 g | |
| HJIMAFKWSKZMBK-UHFFFAOYSA-N | |
| 568.30 | |
| Liquid |
Safety and Handling
TSCA : Yes