missing translation for 'onlineSavingsMsg'
Learn More

Trimethobenzamide Hydrochloride 98.0+%, TCI America™

Catalog No. T35255G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35255G 5 g
T35251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35255G Supplier TCI America Supplier No. T35255G

Chemical Identifiers

CAS 554-92-7
Molecular Formula C21H29ClN2O5
Molecular Weight (g/mol) 424.922
MDL Number MFCD00057999
InChI Key WIIZEEPFHXAUND-UHFFFAOYSA-N
Synonym N-[4-[2-(Dimethylamino)ethoxy]benzyl]-3,4,5-trimethoxybenzamide Hydrochloride
PubChem CID 68385
IUPAC Name N-[[4-[2-(dimethylamino)ethoxy]phenyl]methyl]-3,4,5-trimethoxybenzamide;hydrochloride
SMILES CN(C)CCOC1=CC=C(C=C1)CNC(=O)C2=CC(=C(C(=C2)OC)OC)OC.Cl

Specifications

Melting Point 190°C
Color White
Quantity 5 g
Formula Weight 424.92
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethobenzamide Hydrochloride
TSCA No
Recommended Storage Refrigerator