accessibility menu, dialog, popup

UserName

HClH3COOH3COCH3OHNONCH3CH3

CAS RN 554-92-7

IUPAC Name: hydrogen N-({4-[2-(dimethylamino)ethoxy]phenyl}methyl)-3,4,5-trimethoxybenzamide chloride
Synonyms: hydrogen trimethobenzamide chloride
Molecular Weight (g/mol): 424.92
Molecular Formula: C21H29ClN2O5
InChi Key: WIIZEEPFHXAUND-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=CC(=CC(OC)=C1OC)C(=O)NCC1=CC=C(OCCN(C)C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results