missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimesitylphosphine, 98%
Description
It is used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used as an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 185°C to 188°C |
| Quantity | 1 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 388.54 |
| Percent Purity | 98% |
| Chemical Name or Material | Trimesitylphosphine |
Safety and Handling
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only