missing translation for 'onlineSavingsMsg'
Learn More

Trimesitylphosphine, 98%

Catalog No. AAB2111606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2111606 5 g
AAB2111603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2111606 Supplier Thermo Scientific Chemicals Supplier No. B2111606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23897-15-6 | C27H33P | 388.535 g/mol

It is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 23897-15-6
Molecular Formula C27H33P
Molecular Weight (g/mol) 388.535
MDL Number MFCD00014912
InChI Key IDXDWPWXHTXJMZ-UHFFFAOYSA-N
Synonym trimesitylphosphine, tris 2,4,6-trimethylphenyl phosphine, tris 2,4,6-trimethylphenyl phosphane, phosphine, tris 2,4,6-trimethylphenyl, pubchem6447, acmc-1cs30, phosphine,tris 2,4,6-trimethylphenyl
PubChem CID 2733417
IUPAC Name tris(2,4,6-trimethylphenyl)phosphane
SMILES CC1=CC(=C(C(=C1)C)P(C2=C(C=C(C=C2C)C)C)C3=C(C=C(C=C3C)C)C)C

Specifications

Melting Point 185°C to 188°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 388.54
Percent Purity 98%
Chemical Name or Material Trimesitylphosphine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only