missing translation for 'onlineSavingsMsg'
Learn More

Trimebutine 98.0+%, TCI America™

Supplier:  TCI America T304925G

 View more versions of this product

Catalog No. T304925G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Trimebutine
39133-31-8
C22H29NO5
2-(Dimethylamino)-2-phenylbutyl 3,4,5-Trimethoxybenzoate, 3,4,5-Trimethoxybenzoic Acid 2-(Dimethylamino)-2-phenylbutyl Ester
CCC(COC(=O)C1=CC(=C(C(=C1)OC)OC)OC)(C2=CC=CC=C2)N(C)C
387.476
387.48
25 g
82°C
White
MFCD00133873
LORDFXWUHHSAQU-UHFFFAOYSA-N
[2-(dimethylamino)-2-phenylbutyl] 3,4,5-trimethoxybenzoate
5573
≥98.0% (HPLC,T)
Crystalline Powder

Chemical Identifiers

39133-31-8
387.476
LORDFXWUHHSAQU-UHFFFAOYSA-N
5573
CCC(COC(=O)C1=CC(=C(C(=C1)OC)OC)OC)(C2=CC=CC=C2)N(C)C
C22H29NO5
MFCD00133873
2-(Dimethylamino)-2-phenylbutyl 3,4,5-Trimethoxybenzoate, 3,4,5-Trimethoxybenzoic Acid 2-(Dimethylamino)-2-phenylbutyl Ester
[2-(dimethylamino)-2-phenylbutyl] 3,4,5-trimethoxybenzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions