accessibility menu, dialog, popup

UserName

CAS RN 39133-31-8

IUPAC Name: 2-(dimethylamino)-2-phenylbutyl 3,4,5-trimethoxybenzoate
Synonyms: trimebutine trimebutine maleate
Molecular Weight (g/mol): 387.48
Molecular Formula: C22H29NO5
InChi Key: LORDFXWUHHSAQU-UHFFFAOYNA-N
SMILES: CCC(COC(=O)C1=CC(OC)=C(OC)C(OC)=C1)(N(C)C)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results