missing translation for 'onlineSavingsMsg'
Learn More

Trimebutine 98.0+%, TCI America™

Catalog No. p-7137545
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30495G 5 g
T304925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30495G Supplier TCI America Supplier No. T30495G
Only null left

Chemical Identifiers

CAS 39133-31-8
Molecular Formula C22H29NO5
Molecular Weight (g/mol) 387.476
MDL Number MFCD00133873
InChI Key LORDFXWUHHSAQU-UHFFFAOYSA-N
Synonym 2-(Dimethylamino)-2-phenylbutyl 3,4,5-Trimethoxybenzoate, 3,4,5-Trimethoxybenzoic Acid 2-(Dimethylamino)-2-phenylbutyl Ester
PubChem CID 5573
IUPAC Name [2-(dimethylamino)-2-phenylbutyl] 3,4,5-trimethoxybenzoate
SMILES CCC(COC(=O)C1=CC(=C(C(=C1)OC)OC)OC)(C2=CC=CC=C2)N(C)C

Specifications

Melting Point 82°C
Color White
Formula Weight 387.48
Percent Purity ≥98.0% (HPLC,T)
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Trimebutine
TSCA No
Recommended Storage Refrigerator