missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triisopropylsilylacetylene 95.0+%, TCI America™
Supplier: TCI America T168325ML
Specifications
| Triisopropylsilylacetylene | |
| Yellow | |
| C11H22Si | |
| 25 mL | |
| triisopropylsilyl acetylene, ethynyltriisopropylsilane, triisopropylsilylacetylene, ethynyltris propan-2-yl silane, ethynyl-tri propan-2-yl silane, silane, ethynyltris 1-methylethyl, triisopropylsilylacetylen, acmc-209qzk, ethynyltriisopropyl-silane | |
| CC(C)[Si](C#C)(C(C)C)C(C)C | |
| 182.382 | |
| 182.38 | |
| Liquid |
| 89343-06-6 | |
| 100°C | |
| MFCD00075452 | |
| 1993 | |
| KZGWPHUWNWRTEP-UHFFFAOYSA-N | |
| ethynyl-tri(propan-2-yl)silane | |
| 2734682 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 89343-06-6 | |
| 182.382 | |
| KZGWPHUWNWRTEP-UHFFFAOYSA-N | |
| 2734682 | |
| CC(C)[Si](C#C)(C(C)C)C(C)C |
| C11H22Si | |
| MFCD00075452 | |
| triisopropylsilyl acetylene, ethynyltriisopropylsilane, triisopropylsilylacetylene, ethynyltris propan-2-yl silane, ethynyl-tri propan-2-yl silane, silane, ethynyltris 1-methylethyl, triisopropylsilylacetylen, acmc-209qzk, ethynyltriisopropyl-silane | |
| ethynyl-tri(propan-2-yl)silane |
Safety and Handling
TSCA : No