accessibility menu, dialog, popup

UserName

CAS RN 89343-06-6

IUPAC Name: ethynyltris(propan-2-yl)silane
Synonyms: ethynyltriisopropylsilane
Molecular Weight (g/mol): 182.38
Molecular Formula: C11H22Si
InChi Key: KZGWPHUWNWRTEP-UHFFFAOYSA-N
SMILES: CC(C)[Si](C#C)(C(C)C)C(C)C
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)