missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triisopropylsilylacetylene 95.0+%, TCI America™
$141.67 - $493.97
Chemical Identifiers
| CAS | 89343-06-6 |
|---|---|
| Molecular Formula | C11H22Si |
| Molecular Weight (g/mol) | 182.382 |
| MDL Number | MFCD00075452 |
| InChI Key | KZGWPHUWNWRTEP-UHFFFAOYSA-N |
| Synonym | triisopropylsilyl acetylene, ethynyltriisopropylsilane, triisopropylsilylacetylene, ethynyltris propan-2-yl silane, ethynyl-tri propan-2-yl silane, silane, ethynyltris 1-methylethyl, triisopropylsilylacetylen, acmc-209qzk, ethynyltriisopropyl-silane |
| PubChem CID | 2734682 |
| IUPAC Name | ethynyl-tri(propan-2-yl)silane |
| SMILES | CC(C)[Si](C#C)(C(C)C)C(C)C |
Chemical Identifiers
| 89343-06-6 | |
| 182.382 | |
| KZGWPHUWNWRTEP-UHFFFAOYSA-N | |
| 2734682 | |
| CC(C)[Si](C#C)(C(C)C)C(C)C |
| C11H22Si | |
| MFCD00075452 | |
| triisopropylsilyl acetylene, ethynyltriisopropylsilane, triisopropylsilylacetylene, ethynyltris propan-2-yl silane, ethynyl-tri propan-2-yl silane, silane, ethynyltris 1-methylethyl, triisopropylsilylacetylen, acmc-209qzk, ethynyltriisopropyl-silane | |
| ethynyl-tri(propan-2-yl)silane |
Specifications
| 89343-06-6 | |
| 100°C | |
| MFCD00075452 | |
| 1993 | |
| KZGWPHUWNWRTEP-UHFFFAOYSA-N | |
| ethynyl-tri(propan-2-yl)silane | |
| 2734682 | |
| ≥95.0% (GC) | |
| Triisopropylsilylacetylene |
| Yellow | |
| C11H22Si | |
| 5 mL | |
| triisopropylsilyl acetylene, ethynyltriisopropylsilane, triisopropylsilylacetylene, ethynyltris propan-2-yl silane, ethynyl-tri propan-2-yl silane, silane, ethynyltris 1-methylethyl, triisopropylsilylacetylen, acmc-209qzk, ethynyltriisopropyl-silane | |
| CC(C)[Si](C#C)(C(C)C)C(C)C | |
| 182.382 | |
| 182.38 | |
| Liquid |
Safety and Handling
TSCA : No