missing translation for 'onlineSavingsMsg'
Learn More

Triisopropyl borate, 98+%

CAS: 5419-55-6 | C9H21BO3 | 188.074 g/mol

Supplier:  Thermo Scientific Chemicals A17592AE

 View more versions of this product

Catalog No. AAA17592AE


Only null left
Explore available promotions

Description

Description

Triisopropyl borate is used as reagent in Pd-catalyzed coupling reaction with aryl halides such as Suzuki reaction. It is used as a reagent for the preparation of the boronic acids and esters; as a Lewis acid catalyst and involved in the ortho-borylation of 1-substituted naphthalenes. Furthermore, it plays an important role as a catalyst for the production of resins, waxes, paints and varnishes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triisopropyl borate is used as reagent in Pd-catalyzed coupling reaction with aryl halides such as Suzuki reaction. It is used as a reagent for the preparation of the boronic acids and esters; as a Lewis acid catalyst and involved in the ortho-borylation of 1-substituted naphthalenes. Furthermore, it plays an important role as a catalyst for the production of resins, waxes, paints and varnishes.

Solubility
Miscible with ether, ethanol, isopropanol and benzene.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Decomposes in water.
Specifications

Specifications

Triisopropyl borate
-59°C
139°C to 141°C
Alcohol-like
1.376
MFCD00008872
UN2616
Moisture sensitive
Miscible with ether,ethanol,isopropanol and benzene.
B(OC(C)C)(OC(C)C)OC(C)C
188.074
188.08
5419-55-6
0.815
17°C (62°F)
C9H21BO3
[(CH3)2CHO]3B
100 mL
1701469
triisopropyl borate, isopropyl borate, triisopropoxyborane, triisopropoxyboron, boron isopropoxide, triisopropyl orthoborate, boric acid triisopropyl ester, triisopropoxy borane, boron triisopropoxide, boric acid h3bo3 , tris 1-methylethyl ester
NHDIQVFFNDKAQU-UHFFFAOYSA-N
tripropan-2-yl borate
21531
≥98%

Chemical Identifiers

5419-55-6
188.074
NHDIQVFFNDKAQU-UHFFFAOYSA-N
21531
B(OC(C)C)(OC(C)C)OC(C)C
C9H21BO3
MFCD00008872
triisopropyl borate, isopropyl borate, triisopropoxyborane, triisopropoxyboron, boron isopropoxide, triisopropyl orthoborate, boric acid triisopropyl ester, triisopropoxy borane, boron triisopropoxide, boric acid h3bo3 , tris 1-methylethyl ester
tripropan-2-yl borate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TRIISOPROPYL BORATE

EINECSNumber : 226-529-9

RTECSNumber : ED5950000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only