accessibility menu, dialog, popup

UserName

CH3CH3OBOH3CCH3OH3CCH3

CAS RN 5419-55-6

IUPAC Name: tris(propan-2-yl) borate
Synonyms: triisopropyl borate
Molecular Weight (g/mol): 188.07
Molecular Formula: C9H21BO3
InChi Key: NHDIQVFFNDKAQU-UHFFFAOYSA-N
SMILES: CC(C)OB(OC(C)C)OC(C)C
Boiling Point 
  • (3)
  • (7)
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (7)
  • (3)
  • (7)
  • (4)
  • (6)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (7)