missing translation for 'onlineSavingsMsg'
Learn More

Triisopropyl borate, 98+%

Catalog No. p-7023738
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA17592AE 100 mL
AAA17592AP 500 mL
AAA175920F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA17592AE Supplier Thermo Scientific Chemicals Supplier No. A17592AE
Only null left

CAS: 5419-55-6 | C9H21BO3 | 188.074 g/mol

Triisopropyl borate is used as reagent in Pd-catalyzed coupling reaction with aryl halides such as Suzuki reaction. It is used as a reagent for the preparation of the boronic acids and esters; as a Lewis acid catalyst and involved in the ortho-borylation of 1-substituted naphthalenes. Furthermore, it plays an important role as a catalyst for the production of resins, waxes, paints and varnishes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triisopropyl borate is used as reagent in Pd-catalyzed coupling reaction with aryl halides such as Suzuki reaction. It is used as a reagent for the preparation of the boronic acids and esters; as a Lewis acid catalyst and involved in the ortho-borylation of 1-substituted naphthalenes. Furthermore, it plays an important role as a catalyst for the production of resins, waxes, paints and varnishes.

Solubility
Miscible with ether, ethanol, isopropanol and benzene.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Decomposes in water.

Chemical Identifiers

CAS 5419-55-6
Molecular Formula C9H21BO3
Molecular Weight (g/mol) 188.074
MDL Number MFCD00008872
InChI Key NHDIQVFFNDKAQU-UHFFFAOYSA-N
Synonym triisopropyl borate, isopropyl borate, triisopropoxyborane, triisopropoxyboron, boron isopropoxide, triisopropyl orthoborate, boric acid triisopropyl ester, triisopropoxy borane, boron triisopropoxide, boric acid h3bo3 , tris 1-methylethyl ester
PubChem CID 21531
IUPAC Name tripropan-2-yl borate
SMILES B(OC(C)C)(OC(C)C)OC(C)C

Specifications

Melting Point -59°C
Density 0.815
Boiling Point 139°C to 141°C
Flash Point 17°C (62°F)
Odor Alcohol-like
Refractive Index 1.376
Linear Formula [(CH3)2CHO]3B
Quantity 100 mL
UN Number UN2616
Beilstein 1701469
Sensitivity Moisture sensitive
Solubility Information Miscible with ether,ethanol,isopropanol and benzene.
Formula Weight 188.08
Percent Purity ≥98%
Chemical Name or Material Triisopropyl borate
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TRIISOPROPYL BORATE
EINECSNumber 226-529-9
RTECSNumber ED5950000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only