missing translation for 'onlineSavingsMsg'
Learn More

Triisopropanolamine Borate 98.0+%, TCI America™

Supplier:  TCI America T294325G

 View more versions of this product

Catalog No. T294325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Triisopropanolamine Borate
155°C
135°C
MFCD03791298
triisopropanolamine cyclic borate, 3,7,10-trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, borester 21, triisopropanolamine borate, boric acid, tris 1-amino-2-propyl ester, 2-propanol, 1,1',1-nitrilotri-, cyclic borate, 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo 3.3.3 undecane, 2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, 3,7,10-trimethyl, triisopropanolamineborate
B12OC(CN(CC(O1)C)CC(O2)C)C
199.057
199.06
Crystalline Powder
101-00-8
White-Yellow
C9H18BNO3
25 g
IWKGJTDSJPLUCE-UHFFFAOYSA-N
3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo[3.3.3]undecane
225550
≥98.0% (N)

Chemical Identifiers

101-00-8
199.057
IWKGJTDSJPLUCE-UHFFFAOYSA-N
225550
B12OC(CN(CC(O1)C)CC(O2)C)C
C9H18BNO3
MFCD03791298
triisopropanolamine cyclic borate, 3,7,10-trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, borester 21, triisopropanolamine borate, boric acid, tris 1-amino-2-propyl ester, 2-propanol, 1,1',1-nitrilotri-, cyclic borate, 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo 3.3.3 undecane, 2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, 3,7,10-trimethyl, triisopropanolamineborate
3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo[3.3.3]undecane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : ED5787000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions