accessibility menu, dialog, popup

UserName

H3CNCH3OBOOH3C

CAS RN 101-00-8

IUPAC Name: 3,7,10-trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo[3.3.3]undecane
Synonyms: 3,7,10-trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo[3.3.3]undecane
Molecular Weight (g/mol): 199.06
Molecular Formula: C9H18BNO3
InChi Key: IWKGJTDSJPLUCE-UHFFFAOYNA-N
SMILES: CC1CN2CC(C)OB(O1)OC(C)C2
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)