missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triisopropanolamine Borate 98.0+%, TCI America™
$132.35 - $467.87
Chemical Identifiers
| CAS | 101-00-8 |
|---|---|
| Molecular Formula | C9H18BNO3 |
| Molecular Weight (g/mol) | 199.057 |
| MDL Number | MFCD03791298 |
| InChI Key | IWKGJTDSJPLUCE-UHFFFAOYSA-N |
| Synonym | triisopropanolamine cyclic borate, 3,7,10-trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, borester 21, triisopropanolamine borate, boric acid, tris 1-amino-2-propyl ester, 2-propanol, 1,1',1-nitrilotri-, cyclic borate, 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo 3.3.3 undecane, 2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, 3,7,10-trimethyl, triisopropanolamineborate |
| PubChem CID | 225550 |
| IUPAC Name | 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo[3.3.3]undecane |
| SMILES | B12OC(CN(CC(O1)C)CC(O2)C)C |
Chemical Identifiers
| 101-00-8 | |
| 199.057 | |
| IWKGJTDSJPLUCE-UHFFFAOYSA-N | |
| 225550 | |
| B12OC(CN(CC(O1)C)CC(O2)C)C |
| C9H18BNO3 | |
| MFCD03791298 | |
| triisopropanolamine cyclic borate, 3,7,10-trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, borester 21, triisopropanolamine borate, boric acid, tris 1-amino-2-propyl ester, 2-propanol, 1,1',1-nitrilotri-, cyclic borate, 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo 3.3.3 undecane, 2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, 3,7,10-trimethyl, triisopropanolamineborate | |
| 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo[3.3.3]undecane |
Specifications
| 101-00-8 | |
| White-Yellow | |
| C9H18BNO3 | |
| 5 g | |
| IWKGJTDSJPLUCE-UHFFFAOYSA-N | |
| 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo[3.3.3]undecane | |
| 225550 | |
| ≥98.0% (N) | |
| Triisopropanolamine Borate |
| 155°C | |
| 135°C | |
| MFCD03791298 | |
| triisopropanolamine cyclic borate, 3,7,10-trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, borester 21, triisopropanolamine borate, boric acid, tris 1-amino-2-propyl ester, 2-propanol, 1,1',1-nitrilotri-, cyclic borate, 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo 3.3.3 undecane, 2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, 3,7,10-trimethyl, triisopropanolamineborate | |
| B12OC(CN(CC(O1)C)CC(O2)C)C | |
| 199.057 | |
| 199.06 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : ED5787000
TSCA : No