missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoromethanesulfonanilide 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 67°C |
| Color | White |
| Quantity | 1 g |
| UN Number | 2811 |
| Formula Weight | 225.19 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Trifluoromethanesulfonanilide |
Sécurité et manipulation
| RTECSNumber | PB0482000 |
| TSCA | No |