missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylene Glycol Dimethyl Ether (stabilized with BHT) 99.0+%, TCI America™
$48.46 - $126.76
Chemical Identifiers
| CAS | 112-49-2 |
|---|---|
| Molecular Formula | C8H18O4 |
| Molecular Weight (g/mol) | 178.228 |
| MDL Number | MFCD00008504 |
| InChI Key | YFNKIDBQEZZDLK-UHFFFAOYSA-N |
| Synonym | triglyme, triethylene glycol dimethyl ether, 2,5,8,11-tetraoxadodecane, 1,2-bis 2-methoxyethoxy ethane, glyme 4, ansul ether 161, glyme-3, tegdime, tegdme, unii-32yxg88kk0 |
| PubChem CID | 8189 |
| ChEBI | CHEBI:44842 |
| IUPAC Name | 1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane |
| SMILES | COCCOCCOCCOC |
Chemical Identifiers
| 112-49-2 | |
| 178.228 | |
| YFNKIDBQEZZDLK-UHFFFAOYSA-N | |
| 8189 | |
| 1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane |
| C8H18O4 | |
| MFCD00008504 | |
| triglyme, triethylene glycol dimethyl ether, 2,5,8,11-tetraoxadodecane, 1,2-bis 2-methoxyethoxy ethane, glyme 4, ansul ether 161, glyme-3, tegdime, tegdme, unii-32yxg88kk0 | |
| CHEBI:44842 | |
| COCCOCCOCCOC |
Specifications
| 112-49-2 | |
| Colorless | |
| C8H18O4 | |
| 25 g | |
| YFNKIDBQEZZDLK-UHFFFAOYSA-N | |
| 1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane | |
| 8189 | |
| 178.23 | |
| Liquid |
| -45°C | |
| 216°C | |
| MFCD00008504 | |
| triglyme, triethylene glycol dimethyl ether, 2,5,8,11-tetraoxadodecane, 1,2-bis 2-methoxyethoxy ethane, glyme 4, ansul ether 161, glyme-3, tegdime, tegdme, unii-32yxg88kk0 | |
| COCCOCCOCCOC | |
| 178.228 | |
| CHEBI:44842 | |
| ≥99.0% (GC) | |
| Triethylene Glycol Dimethyl Ether (stabilized with BHT) |
Safety and Handling
EINECSNumber : (7)-1321
RTECSNumber : XF0665000
TSCA : Yes