accessibility menu, dialog, popup

UserName

CAS RN 112-49-2

IUPAC Name: 2,5,8,11-tetraoxadodecane
Synonyms: triglyme (tegdme) tegdme glyme-3 glyme 4 ansul ether 161 tegdime
Molecular Weight (g/mol): 178.23
Molecular Formula: C8H18O4
InChi Key: YFNKIDBQEZZDLK-UHFFFAOYSA-N
SMILES: COCCOCCOCCOC
Boiling Point 
  • (3)
  • (5)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (3)
  • (5)
  • (5)
  • (3)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)