missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylene Glycol Dimethyl Ether (stabilized with BHT) 99.0+%, TCI America™
Supplier: TCI America B0496500G
Specifications
| Triethylene Glycol Dimethyl Ether (stabilized with BHT) | |
| -45°C | |
| 216°C | |
| MFCD00008504 | |
| triglyme, triethylene glycol dimethyl ether, 2,5,8,11-tetraoxadodecane, 1,2-bis 2-methoxyethoxy ethane, glyme 4, ansul ether 161, glyme-3, tegdime, tegdme, unii-32yxg88kk0 | |
| COCCOCCOCCOC | |
| 178.228 | |
| CHEBI:44842 | |
| ≥99.0% (GC) |
| 112-49-2 | |
| Colorless | |
| C8H18O4 | |
| 500 g | |
| YFNKIDBQEZZDLK-UHFFFAOYSA-N | |
| 1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane | |
| 8189 | |
| 178.23 | |
| Liquid |
Chemical Identifiers
| 112-49-2 | |
| 178.228 | |
| YFNKIDBQEZZDLK-UHFFFAOYSA-N | |
| 8189 | |
| 1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane |
| C8H18O4 | |
| MFCD00008504 | |
| triglyme, triethylene glycol dimethyl ether, 2,5,8,11-tetraoxadodecane, 1,2-bis 2-methoxyethoxy ethane, glyme 4, ansul ether 161, glyme-3, tegdime, tegdme, unii-32yxg88kk0 | |
| CHEBI:44842 | |
| COCCOCCOCCOC |
Safety and Handling
EINECSNumber : (7)-1321
RTECSNumber : XF0665000
TSCA : Yes