missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylborane, 1M solution in THF, AcroSeal™
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
Specifications
Specifications
| Color | Amber to Colorless |
| Density | 0.8600g/mL |
| Flash Point | −17°C |
| Packaging | AcroSeal™ Glass bottle |
| Linear Formula | (C2H5)3B |
| Quantity | 100 mL |
| Fieser | 09,482; 10,416; 15,333; 16,160; 17,363 |
| Specific Gravity | 0.86 |
| Merck Index | 15, 9833 |
| Formula Weight | 98 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Highly flammable liquid and vapour. Harmful if swallowed. Harmful if inhaled. Causes severe skin burns and eye damage. May cause respiratory irritation. May cause drowsiness or dizziness. Suspect |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye p |
| EINECSNumber | 202-620-9 |
RUO – Research Use Only