accessibility menu, dialog, popup

UserName

CAS RN 97-94-9

IUPAC Name: triethylborane
Synonyms: triethylborane
Molecular Weight (g/mol): 98
Molecular Formula: C6H15B
InChi Key: LALRXNPLTWZJIJ-UHFFFAOYSA-N
SMILES: CCB(CC)CC
Quantity 
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (3)