missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethyl Orthobenzoate 97.0+%, TCI America™
$75.08 - $246.43
Chemical Identifiers
| CAS | 1663-61-2 |
|---|---|
| Molecular Formula | C13H20O3 |
| Molecular Weight (g/mol) | 224.3 |
| MDL Number | MFCD00009222 |
| InChI Key | BQFPCTXLBRVFJL-UHFFFAOYSA-N |
| Synonym | triethoxymethyl benzene, triethyl orthobenzoate, ethyl orthobenzoate, benzene, triethoxymethyl, triethylorthobenzoate, orthobenzoic acid, triethyl ester, orthobenzoic acid triethyl ester, triethoxymethyl benzol, triethoxyphenylmethane |
| PubChem CID | 74268 |
| IUPAC Name | triethoxymethylbenzene |
| SMILES | CCOC(C1=CC=CC=C1)(OCC)OCC |
Chemical Identifiers
| 1663-61-2 | |
| 224.3 | |
| BQFPCTXLBRVFJL-UHFFFAOYSA-N | |
| 74268 | |
| CCOC(C1=CC=CC=C1)(OCC)OCC |
| C13H20O3 | |
| MFCD00009222 | |
| triethoxymethyl benzene, triethyl orthobenzoate, ethyl orthobenzoate, benzene, triethoxymethyl, triethylorthobenzoate, orthobenzoic acid, triethyl ester, orthobenzoic acid triethyl ester, triethoxymethyl benzol, triethoxyphenylmethane | |
| triethoxymethylbenzene |
Specifications
| 1663-61-2 | |
| 240°C | |
| MFCD00009222 | |
| triethoxymethyl benzene, triethyl orthobenzoate, ethyl orthobenzoate, benzene, triethoxymethyl, triethylorthobenzoate, orthobenzoic acid, triethyl ester, orthobenzoic acid triethyl ester, triethoxymethyl benzol, triethoxyphenylmethane | |
| CCOC(C1=CC=CC=C1)(OCC)OCC | |
| 224.3 | |
| 224.30 | |
| Liquid |
| Colorless | |
| C13H20O3 | |
| 5 g | |
| BQFPCTXLBRVFJL-UHFFFAOYSA-N | |
| triethoxymethylbenzene | |
| 74268 | |
| ≥97.0% (GC) | |
| Triethyl Orthobenzoate |
Safety and Handling
TSCA : Yes