accessibility menu, dialog, popup

UserName

CAS RN 1663-61-2

IUPAC Name: (triethoxymethyl)benzene
Synonyms: (triethoxymethyl)benzene
Molecular Weight (g/mol): 224.3
Molecular Formula: C13H20O3
InChi Key: BQFPCTXLBRVFJL-UHFFFAOYSA-N
SMILES: CCOC(OCC)(OCC)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)