missing translation for 'onlineSavingsMsg'
Learn More

Triethyl orthobenzoate, 97%

Catalog No. p-3213274
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC378760250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC378760250 Supplier Thermo Scientific Chemicals Supplier No. 378760250

CAS: 1663-61-2 | C13H20O3 | 224.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent

Chemical Identifiers

CAS 1663-61-2
Molecular Formula C13H20O3
Molecular Weight (g/mol) 224.3
MDL Number MFCD00009222
InChI Key BQFPCTXLBRVFJL-UHFFFAOYSA-N
Synonym triethoxymethyl benzene, triethyl orthobenzoate, ethyl orthobenzoate, benzene, triethoxymethyl, triethylorthobenzoate, orthobenzoic acid, triethyl ester, orthobenzoic acid triethyl ester, triethoxymethyl benzol, triethoxyphenylmethane
PubChem CID 74268
IUPAC Name triethoxymethylbenzene
SMILES CCOC(C1=CC=CC=C1)(OCC)OCC

Specifications

Color Colorless
Density 0.9910g/mL
Boiling Point 239°C to 241°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4710 to 1.474
Linear Formula C6H5C(OCH2CH3)3
Quantity 25 g
Specific Gravity 0.991
Solubility Information Solubility in water: hydrolysis
Formula Weight 224.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Triethyl orthobenzoate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-771-3

RUO – Research Use Only