missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tricyclo[5.2.1.0(2,6)]decan-8-one 95.0+%, TCI America™
Supplier: TCI America T132125ML
Specifications
| Tricyclo[5.2.1.0(2,6)]decan-8-one | |
| Colorless | |
| 132°C | |
| MFCD00168034 | |
| OMIDXVJKZCPKEI-UHFFFAOYSA-N | |
| 150.221 | |
| 150.22 | |
| Liquid |
| 13380-94-4 | |
| 7 (3g/L H2O soln. 20°C) | |
| C10H14O | |
| 25 mL | |
| C1CC2C(C1)C3CC2CC3=O | |
| 96509 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 13380-94-4 | |
| 150.221 | |
| OMIDXVJKZCPKEI-UHFFFAOYSA-N | |
| C1CC2C(C1)C3CC2CC3=O |
| C10H14O | |
| MFCD00168034 | |
| 96509 |
Safety and Handling
RTECSNumber : PC0350000
TSCA : Yes