missing translation for 'onlineSavingsMsg'
Learn More

Tricyclo[5.2.1.0(2,6)]decan-8-one 95.0+%, TCI America™

$111.84 - $754.94

Chemical Identifiers

CAS 13380-94-4
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00168034
InChI Key OMIDXVJKZCPKEI-UHFFFAOYSA-N
PubChem CID 96509
SMILES C1CC2C(C1)C3CC2CC3=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T132125ML
View Documents
TCI America
T132125ML
25 mL
Each for $111.84
Only null left
 
T1321500ML
View Documents
TCI America
T1321500ML
500 mL
Each for $754.94
Only null left
 
Description

Description

Specifications

Chemical Identifiers

13380-94-4
150.221
OMIDXVJKZCPKEI-UHFFFAOYSA-N
C1CC2C(C1)C3CC2CC3=O
C10H14O
MFCD00168034
96509

Specifications

13380-94-4
7 (3g/L H2O soln. 20°C)
C10H14O
25 mL
C1CC2C(C1)C3CC2CC3=O
96509
≥95.0% (GC)
Tricyclo[5.2.1.0(2,6)]decan-8-one
Colorless
132°C
MFCD00168034
OMIDXVJKZCPKEI-UHFFFAOYSA-N
150.221
150.22
Liquid
Videos
Safety and Handling

Safety and Handling

RTECSNumber : PC0350000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'