missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Tricine, ULTROL™ Grade, Calbiochem™,
A zwitterionic buffer useful in the pH range of 7.4 to 8.8
$131.00 - $861.00
Chemical Identifiers
| CAS | 4-1-5704 |
|---|---|
| Molecular Formula | C6H13NO5 |
| Molecular Weight (g/mol) | 179.17 |
| InChI Key | SEQKRHFRPICQDD-UHFFFAOYSA-N |
| Synonym | N-[tris(Hydroxymethyl)methyl]glycine |
| PubChem CID | 79784 |
| ChEBI | CHEBI:39063 |
| IUPAC Name | 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid |
| SMILES | C(C(=O)O)NC(CO)(CO)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
39468100GM
|
MilliporeSigma™
39468100GM |
100 g |
N/A
|
|
|||||
|
394681KG
|
MilliporeSigma™
394681KG |
1 kg |
N/A
|
|
|||||
Chemical Identifiers
| 4-1-5704 | |
| 179.17 | |
| N-[tris(Hydroxymethyl)methyl]glycine | |
| CHEBI:39063 | |
| C(C(=O)O)NC(CO)(CO)CO |
| C6H13NO5 | |
| SEQKRHFRPICQDD-UHFFFAOYSA-N | |
| 79784 | |
| 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid |
Specifications
| 4-1-5704 | |
| ≥99.0% | |
| N-[tris(Hydroxymethyl)methyl]glycine | |
| SEQKRHFRPICQDD-UHFFFAOYSA-N | |
| 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid | |
| 79784 | |
| 179.2 | |
| zwitterionic buffer |
| 100 g | |
| C6H13NO5 | |
| Not available | |
| C(C(=O)O)NC(CO)(CO)CO | |
| 179.17 | |
| CHEBI:39063 | |
| by titration |
Safety and Handling
Recommended Storage : 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).