missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Tricine, ULTROL™ Grade, Calbiochem™,

Catalog No. 39468100GM
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39468100GM 100 g
394681KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 39468100GM Supplier MilliporeSigma™ Supplier No. 39468100GM
Only null left

A zwitterionic buffer useful in the pH range of 7.4 to 8.8

Chemical Identifiers

CAS 4-1-5704
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
Synonym N-[tris(Hydroxymethyl)methyl]glycine
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO

Specifications

Quantity 100 g
Assay Percent Range ≥99.0%
Solubility Information Not available
Formula Weight 179.2
Grade by titration
Chemical Name or Material zwitterionic buffer
Recommended Storage 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).