missing translation for 'onlineSavingsMsg'
Learn More

Tricine, Electrophoresis Grade, MP Biomedicals

Catalog No. ICN807413
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN807413 100 g
ICN807416 250 g
ICN807420 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN807413 Supplier MP Biomedicals Inc Supplier No. 04807413
Only null left

5704-04-1 Purity: >>>99% Heavy metals (as Pb): <<10 ppm pKa at 25°C = 8.1 Biological buffer

Chemical Identifiers

CAS 4-1-5704
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO

Specifications

Quantity 100 g
Grade Electrophoresis
Chemical Name or Material Tricine