missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tricine, Electrophoresis Grade, 99%
CAS: 5704-04-1 | C6H13NO5 | 179.17 g/mol
$160.64 - $549.01
Chemical Identifiers
| CAS | 4-1-5704 |
|---|---|
| Molecular Formula | C6H13NO5 |
| Molecular Weight (g/mol) | 179.17 |
| MDL Number | MFCD00004277 |
| InChI Key | SEQKRHFRPICQDD-UHFFFAOYSA-N |
| Synonym | N-(Trishydroxymethylmethyl)glycine |
| PubChem CID | 79784 |
| ChEBI | CHEBI:39063 |
| IUPAC Name | 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid |
| SMILES | C(C(=O)O)NC(CO)(CO)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6443722
|
Thermo Scientific Chemicals
J6443722 |
100 g |
Each for $160.64
|
|
|||||
|
AAJ6443736
|
Thermo Scientific Chemicals
J6443736 |
500 g |
Each for $549.01
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4-1-5704 | |
| 179.17 | |
| SEQKRHFRPICQDD-UHFFFAOYSA-N | |
| 79784 | |
| 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid |
| C6H13NO5 | |
| MFCD00004277 | |
| N-(Trishydroxymethylmethyl)glycine | |
| CHEBI:39063 | |
| C(C(=O)O)NC(CO)(CO)CO |
Specifications
| ∽183°C (decomposition) | |
| White | |
| 100 g | |
| (HOCH2)3CNHCH2CO2H | |
| 1937804 | |
| N-(Trishydroxymethylmethyl)glycine | |
| C(C(=O)O)NC(CO)(CO)CO | |
| 179.17 | |
| CHEBI:39063 | |
| 99% | |
| Tricine |
| 4-1-5704 | |
| Powder | |
| C6H13NO5 | |
| MFCD00004277 | |
| 14,9651 | |
| SEQKRHFRPICQDD-UHFFFAOYSA-N | |
| 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid | |
| 79784 | |
| 179.17 | |
| Electrophoresis |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 227-193-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only