missing translation for 'onlineSavingsMsg'
Learn More

Tricine, Electrophoresis Grade, 99%

Catalog No. AAJ6443736
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6443736 500 g
AAJ6443722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6443736 Supplier Thermo Scientific Chemicals Supplier No. J6443736
Only null left

CAS: 5704-04-1 | C6H13NO5 | 179.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4-1-5704
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
Synonym N-(Trishydroxymethylmethyl)glycine
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO

Specifications

Melting Point ∽183°C (decomposition)
Color White
Physical Form Powder
Quantity 500 g
Linear Formula (HOCH2)3CNHCH2CO2H
Beilstein 1937804
Merck Index 14,9651
Formula Weight 179.17
Percent Purity 99%
Grade Electrophoresis
Chemical Name or Material Tricine
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 227-193-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only