missing translation for 'onlineSavingsMsg'
Learn More

Tricine-buffered saline (5X), pH 7.0, Thermo Scientific™

Catalog No. AAJ60811AK
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60811AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60811AK Supplier Thermo Scientific Chemicals Supplier No. J60811AK
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 4-1-5704
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO

Specifications

Color Colorless
Concentration 5X
pH 7.0
Physical Form Liquid
Quantity 250 mL
Odor Odorless
Chemical Name or Material Tricine-buffered saline
TSCA Yes
Recommended Storage Ambient temperatures