missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tricine Buffer, MP Biomedicals
$72.88 - $264.43
Chemical Identifiers
| CAS | 4-1-5704 |
|---|---|
| Molecular Formula | C6H13NO5 |
| Molecular Weight (g/mol) | 179.17 |
| MDL Number | MFCD00004277 |
| InChI Key | SEQKRHFRPICQDD-UHFFFAOYSA-N |
| Synonym | N-Tris-[hydroxymethyl]methylglycine, N-[2-Hydroxy-1, 1-bis(hydroxymethyl) ethyl] glycine |
| PubChem CID | 79784 |
| ChEBI | CHEBI:39063 |
| IUPAC Name | 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid |
| SMILES | C(C(=O)O)NC(CO)(CO)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10311225
|
MP Biomedicals Inc
0210311225 |
25 g |
Each for $72.88
|
|
|||||
|
ICN10311280
|
MP Biomedicals Inc
0210311280 |
100 g |
Each for $123.66
|
|
|||||
|
ICN10311283
|
MP Biomedicals Inc
0210311283 |
250 g |
Each for $264.43
|
|
|||||
Chemical Identifiers
| 4-1-5704 | |
| 179.17 | |
| SEQKRHFRPICQDD-UHFFFAOYSA-N | |
| 79784 | |
| 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid |
| C6H13NO5 | |
| MFCD00004277 | |
| N-Tris-[hydroxymethyl]methylglycine, N-[2-Hydroxy-1, 1-bis(hydroxymethyl) ethyl] glycine | |
| CHEBI:39063 | |
| C(C(=O)O)NC(CO)(CO)CO |
Specifications
| 4-1-5704 | |
| Powder | |
| C6H13NO5 | |
| N-[2-Hydroxy-1, 1-bis(hydroxymethyl) ethyl] glycine, N-Tris-[hydroxymethyl]methylglycine | |
| C(C(=O)O)NC(CO)(CO)CO | |
| 179.17 | |
| CHEBI:39063 | |
| Tricine Buffer, Cell Biology and Analysis, Molecular Biology |
| White | |
| 25 g | |
| MFCD00004277 | |
| SEQKRHFRPICQDD-UHFFFAOYSA-N | |
| 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid | |
| 79784 | |
| 179.2 |
Safety and Handling
EINECSNumber : 227-193-6
Recommended Storage : Store at room temperature (15° to 30°C).