missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tricine Buffer, MP Biomedicals
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Quantity | 25 g |
| Formula Weight | 179.2 |
| Chemical Name or Material | Tricine Buffer, Cell Biology and Analysis, Molecular Biology |
Safety and Handling
| EINECSNumber | 227-193-6 |
| Recommended Storage | Store at room temperature (15° to 30°C). |