missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Tricine, 99+%, for biochemistry
CAS: 5704-04-1 | C6H13NO5 | 179.17 g/mol
$71.68 - $3887.72
Chemical Identifiers
| CAS | 4-1-5704 |
|---|---|
| Molecular Formula | C6H13NO5 |
| Molecular Weight (g/mol) | 179.17 |
| MDL Number | MFCD00004277 |
| InChI Key | SEQKRHFRPICQDD-UHFFFAOYSA-N |
| Synonym | N-[Tris(hydroxymethyl)methyl]glycine, N-(2-Hydroxy-1, 1-bis(hydroxymethyl)ethyl)glycine |
| PubChem CID | 79784 |
| ChEBI | CHEBI:39063 |
| IUPAC Name | 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid |
| SMILES | C(C(=O)O)NC(CO)(CO)CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC327830250
|
Thermo Scientific Chemicals
327830250 |
25 g | Plastic bottle |
Each for $71.68
|
|
||||
|
AC327831000
|
Thermo Scientific Chemicals
327831000 |
100 g | Plastic bottle |
Each for $159.57
|
|
||||
|
AC327830010
|
Thermo Scientific Chemicals
327830010 |
1 kg | Plastic bottle |
Each for $932.67
|
|
||||
|
AC327830050
|
Thermo Scientific Chemicals
327830050 |
5 kg | Plastic drum |
Each for $3,887.72
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4-1-5704 | |
| 179.17 | |
| SEQKRHFRPICQDD-UHFFFAOYSA-N | |
| 79784 | |
| 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid |
| C6H13NO5 | |
| MFCD00004277 | |
| N-[Tris(hydroxymethyl)methyl]glycine, N-(2-Hydroxy-1, 1-bis(hydroxymethyl)ethyl)glycine | |
| CHEBI:39063 | |
| C(C(=O)O)NC(CO)(CO)CO |
Specifications
| 0.02 max. at 280nm | |
| 180.0°C to 184.0°C | |
| White | |
| Crystalline Powder | |
| 99+% | |
| C6H13NO5 | |
| MFCD00004277 | |
| N-[Tris(hydroxymethyl)methyl]glycine, N-(2-Hydroxy-1, 1-bis(hydroxymethyl)ethyl)glycine | |
| SEQKRHFRPICQDD-UHFFFAOYSA-N | |
| 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid | |
| 79784 | |
| 179.17 | |
| Biochemical |
| (0.1M aq. soln.) | |
| 4-1-5704 | |
| 4.6 to 5.6 (1% aq. soln. at 25°C) | |
| 25 g | |
| Authentic | |
| (HOCH2)3CNHCH2CO2H | |
| Plastic bottle | |
| Solubility in water: soluble. | |
| C(C(=O)O)NC(CO)(CO)CO | |
| 179.17 | |
| CHEBI:39063 | |
| ≥99% | |
| Tricine |
Safety and Handling
Causes serious eye irritation, May cause respiratory irritation, Causes skin irritation
Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
RUO – Research Use Only