missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Tricine, 99+%, for biochemistry
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Absorbance | 0.02 max. at 280nm |
| Absorbance Conditions | (0.1M aq. soln.) |
| Melting Point | 180.0°C to 184.0°C |
| Color | White |
| pH | 4.6 to 5.6 (1% aq. soln. at 25°C) |
| Physical Form | Crystalline Powder |
| Quantity | 100 g |
| Assay Percent Range | 99+% |
| Infrared Spectrum | Authentic |
| Linear Formula | (HOCH2)3CNHCH2CO2H |
| Show More |
Safety and Handling
| Hazard Category | Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2) |
| Hazard Statement | Causes serious eye irritation, May cause respiratory irritation, Causes skin irritation |
RUO – Research Use Only