missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tricine, 99+%, for biochemistry

Catalog No. AC327830250
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC327830250 25 g Plastic bottle
AC327831000 100 g Plastic bottle
AC327830010 1 kg Plastic bottle
AC327830050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC327830250 Supplier Thermo Scientific Chemicals Supplier No. 327830250
Only null left

CAS: 5704-04-1 | C6H13NO5 | 179.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4-1-5704
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
Synonym N-[Tris(hydroxymethyl)methyl]glycine, N-(2-Hydroxy-1, 1-bis(hydroxymethyl)ethyl)glycine
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO

Specifications

Absorbance 0.02 max. at 280nm
Absorbance Conditions (0.1M aq. soln.)
Melting Point 180.0°C to 184.0°C
Color White
pH 4.6 to 5.6 (1% aq. soln. at 25°C)
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Linear Formula (HOCH2)3CNHCH2CO2H
Packaging Plastic bottle
Solubility Information Solubility in water: soluble.
Formula Weight 179.17
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material Tricine
Show More Show Less
Hazard Category Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
Hazard Statement Causes serious eye irritation, May cause respiratory irritation, Causes skin irritation

RUO – Research Use Only