Learn More
Tricine, 98+%
CAS: 5704-04-1 | C6H13NO5 | 179.17 g/mol
Supplier: Thermo Scientific Chemicals A1469522
Description
Good's buffers Tricine is used as an electrophoresis buffer and involved in the separation of low molecular weight proteins and peptides. It operates in the pH range between 7.4 and 8.8. It acts as an effective scavenger of hydroxyl radicals during the study of radiation-induced membrane damage. It is also used in resuspension of cell pellets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGood′s buffers Tricine is used as an electrophoresis buffer and involved in the separation of low molecular weight proteins and peptides. It operates in the pH range between 7.4 and 8.8. It acts as an effective scavenger of hydroxyl radicals during the study of radiation-induced membrane damage. It is also used in resuspension of cell pellets.
Solubility
Soluble in water.
Notes
pH range: 7.8-8.8Incompatible with strong oxidizing agents.
Specifications
| Tricine | |
| 4-1-5704 | |
| C6H13NO5 | |
| 1937804 | |
| N-(Trishydroxymethylmethyl)glycine | |
| SEQKRHFRPICQDD-UHFFFAOYSA-N | |
| 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid | |
| 79784 | |
| 179.17 |
| ∼183°C (decomposition) | |
| 100 g | |
| MFCD00004277 | |
| 14,9651 | |
| Soluble in water. | |
| C(C(=O)O)NC(CO)(CO)CO | |
| 179.17 | |
| CHEBI:39063 | |
| ≥98% |
Chemical Identifiers
| 4-1-5704 | |
| 179.17 | |
| SEQKRHFRPICQDD-UHFFFAOYSA-N | |
| 79784 | |
| 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid |
| C6H13NO5 | |
| MFCD00004277 | |
| N-(Trishydroxymethylmethyl)glycine | |
| CHEBI:39063 | |
| C(C(=O)O)NC(CO)(CO)CO |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 227-193-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only