missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Tricine, 0.5M buffer soln., pH 8.0
CAS: 5704-04-1 | C6H13NO5 | 179.17 g/mol
$185.36 - $284.22
Chemical Identifiers
| CAS | 4-1-5704 |
|---|---|
| Molecular Formula | C6H13NO5 |
| Molecular Weight (g/mol) | 179.17 |
| MDL Number | MFCD00004277 |
| InChI Key | SEQKRHFRPICQDD-UHFFFAOYSA-N |
| PubChem CID | 79784 |
| ChEBI | CHEBI:39063 |
| IUPAC Name | 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid |
| SMILES | C(C(=O)O)NC(CO)(CO)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ62983AE
|
Thermo Scientific Chemicals
J62983AE |
100 mL |
Each for $185.36
|
|
|||||
|
AAJ62983AK
|
Thermo Scientific Chemicals
J62983AK |
250 mL |
Each for $284.22
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4-1-5704 | |
| 179.17 | |
| SEQKRHFRPICQDD-UHFFFAOYSA-N | |
| CHEBI:39063 | |
| C(C(=O)O)NC(CO)(CO)CO |
| C6H13NO5 | |
| MFCD00004277 | |
| 79784 | |
| 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid |
Specifications
| 4-1-5704 | |
| 0.5 M | |
| Liquid | |
| C6H13NO5 | |
| SEQKRHFRPICQDD-UHFFFAOYSA-N | |
| 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid | |
| 79784 | |
| Tricine buffer soln. |
| Colorless | |
| 8.0 | |
| 100 mL | |
| MFCD00004277 | |
| C(C(=O)O)NC(CO)(CO)CO | |
| 179.17 | |
| CHEBI:39063 |
Safety and Handling
H335
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only